Risperidone for system suitability
Synonym(s):
Risperidone, R 64,766
Empirical Formula (Hill Notation):
C23H27FN4O2
CAS Number:
Molecular Weight:
410.48
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing