Skip to Content
Merck
Riociguat ≥98% (HPLC)

Riociguat

Riociguat ≥98% (HPLC)
Synonym(s):
N-[4,6-Diamino-2-[1-[(2-fluorophenyl)methyl]-1H-pyrazolo[3,4-b]pyridin-3-yl]-5-pyrimidinyl]-N-methylcarbamic acid methyl ester, BAY 63-2521, BAY 632521, Methyl [4,6-diamino-2-[1-(2-fluorobenzyl)-1H-pyrazolo[3,4-b]pyridin-3-yl]pyrimidin-5-yl]methylcarbamate, N-[4,6-Diamino-2-[1-[(2-fluorophenyl)methyl]-1H-pyrazolo[3,4-b]pyridin-3-yl]-5-pyrimidinyl]-N-methylcarbamic acid methyl ester
Empirical Formula (Hill Notation):
C20H19FN8O2
CAS Number:
Molecular Weight:
422.42

Products

Brand
Color
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type