Skip to Content
Merck
vibrant-m

Residual Solvent Class 1 - 1,2-Dichloroethane

vibrant-m
Synonym(s):
1,2–Dichloroethane solution
Empirical Formula (Hill Notation):
C2H4Cl2
CAS Number:
Molecular Weight:
98.96

Products

Formula Weight
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1