Skip to Content
Merck
Repaglinide Related Compound E pharmaceutical secondary standard, certified reference material

Repaglinide Related Compound E

Repaglinide Related Compound E pharmaceutical secondary standard, certified reference material
Synonym(s):
2-Ethoxy-4-{2-{{(1R)-3-methyl-1-[2-(piperidin-1-yl)phenyl]butyl}amino}-2-oxoethyl}benzoic acid, (R)-(-)-Repaglinide
Empirical Formula (Hill Notation):
C27H36N2O4
CAS Number:
Molecular Weight:
452.59

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade