![(R)-4-Benzyl-2-oxazolidinone 99%](/deepweb/assets/sigmaaldrich/product/structures/282/837/4df681b0-db3a-46f8-a903-20d92825dc00/640/4df681b0-db3a-46f8-a903-20d92825dc00.png)
(R)-4-Benzyl-2-oxazolidinone
![(R)-4-Benzyl-2-oxazolidinone 99%](/deepweb/assets/sigmaaldrich/product/structures/282/837/4df681b0-db3a-46f8-a903-20d92825dc00/640/4df681b0-db3a-46f8-a903-20d92825dc00.png)
Empirical Formula (Hill Notation):
C10H11NO2
CAS Number:
Molecular Weight:
177.20
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing