(R)-1-[(SP)-2-(Diphenylphosphino)ferrocenyl]ethyldi-tert-butylphosphine
Synonym(s):
(2R)-1-[(1R)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(diphenylphosphino)ferrocene (acc to CAS), Josiphos SL-J002-1
Empirical Formula (Hill Notation):
C32H40FeP2
CAS Number:
Molecular Weight:
542.45
Products
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing