Skip to Content
Merck
Propylbenzene analytical standard

Propylbenzene

Propylbenzene analytical standard
Synonym(s):
1-Phenylpropane
Linear Formula:
C6H5CH2CH2CH3
CAS Number:
Molecular Weight:
120.19
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing