Skip to Content
Merck
Propofol Related Compound C pharmaceutical secondary standard, certified reference material

Propofol Related Compound C

Propofol Related Compound C pharmaceutical secondary standard, certified reference material
Synonym(s):
2,6-Diisopropylphenyl isopropyl ether, 2-(1-Methylethoxy)-1,3-bis(1-methylethyl)benzene, Propofol isopropyl ether
Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35

Products

Brand
Feature
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade