Skip to Content
Merck
Potassium phosphate monobasic suitable for cell culture, Redi-Dri™, ≥99.0%, free-flowing, suitable for insect cell culture, suitable for plant cell culture

Potassium phosphate monobasic

Potassium phosphate monobasic suitable for cell culture, Redi-Dri™, ≥99.0%, free-flowing, suitable for insect cell culture, suitable for plant cell culture
Synonym(s):
Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate, Potassium phosphate monobasic
Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-18 of 18
Compare
Product Number
Description
Pricing