Skip to Content
Merck
Potassium indigotrisulfonate ozone-scavenging reagent

Potassium indigotrisulfonate

Potassium indigotrisulfonate ozone-scavenging reagent
Synonym(s):
5,5′,7-Indigotrisulfonic acid tripotassium salt, Indigo-5,5′,7-trisulfonic acid tripotassium salt, Potassium indigotrisulfonate, Tripotassium indigo-5,5′,7-trisulfonate, tri-Potassium indigo-5,5′,7-trisulfonate
Empirical Formula (Hill Notation):
C16H7K3N2O11S3
CAS Number:
Molecular Weight:
616.72

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing