Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Polyisobutylene
Linear Formula:
[CH
2
C(CH
3
)
2
]
n
CAS Number:
9003-27-4
Products
Brand
Sigma-Aldrich (2)
Color
Manufacturer
Physical Form
Format
Quality Segment
Application
Polymer Composition
Polymer Type
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
181455
Polyisobutylene,
average M
w
~500,000, average M
n
~200,000 by GPC/MALLS, average M
v
~420,000
Expand
View Pricing
181463
Polyisobutylene,
average M
w
~1,000,000, average M
n
~600,000 by GPC/MALLS, average M
v
~1,200,000
Expand
View Pricing
1546966
Polyisobutylene,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing