Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Polycarbonate
Products
Color
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-20 of 118
Page 1 of 6
Page 1 of 6
Compare
Product Number
Description
Pricing
GF57272485
Polycarbonate,
(PC), sheet, thickness 2 mm, size 150 × 150 mm, clear
Expand
View Pricing
GF65553598
Polycarbonate,
(PC), granule, 3 mm nominal granule size, weight 100 g, additives heat stabilizer only
Expand
View Pricing
GF61784242
Polycarbonate,
(PC), granule, 3 mm nominal granule size, weight 1000 g, additives heat stabilizer only
Expand
View Pricing
GF60306689
Polycarbonate,
(PC), sheet, thickness 0.5 mm, size 500 × 1000 mm, grade makrofol de 1-4
Expand
View Pricing
GF59060875
Polycarbonate,
(PC), sheet, thickness 2 mm, size 300 × 300 mm, clear
Expand
View Pricing
GF43917408
Polycarbonate,
(PC), sheet, thickness 0.5 mm, size 100 × 100 mm, grade makrofol de 1-4
Expand
View Pricing
GF46105411
Polycarbonate,
(PC), film, thickness 0.25 mm, size 600 × 600 mm, clear
Expand
View Pricing
GF32517020
Polycarbonate,
(PC), sheet, thickness 1 mm, size 150 × 150 mm, clear
Expand
View Pricing
GF23031010
Polycarbonate,
(PC), granule, 3 mm nominal granule size, weight 2000 g, additives heat stabilizer only
Expand
View Pricing
GF82609566
Polycarbonate,
(PC), sheet, thickness 2 mm, size 500 × 500 mm, clear
Expand
View Pricing
GF86768847
Polycarbonate,
(PC), film, thickness 0.25 mm, size 300 × 300 mm, clear
Expand
View Pricing
GF77460475
Polycarbonate,
(PC), sheet, thickness 2 mm, size 600 × 600 mm, clear
Expand
View Pricing
GF80430027
Polycarbonate,
(PC), tube, outside diameter 20mm, L 500 mm, wall thickness 2 mm
Expand
View Pricing
GF01715217
Polycarbonate,
(PC), film, thickness 0.002 mm, L 2 m
Expand
View Pricing
GF64192901
Polycarbonate,
(PC), film, thickness 0.175 mm, size 300 × 300 mm, clear
Expand
View Pricing
GF52971363
Polycarbonate,
(PC), sheet, thickness 5 mm, size 300 × 300 mm, clear
Expand
View Pricing
GF21421088
Polycarbonate,
(PC), sheet, thickness 5 mm, size 500 × 500 mm, clear
Expand
View Pricing
GF49028632
Polycarbonate,
(PC), sheet, thickness 1 mm, size 600 × 600 mm, clear
Expand
View Pricing
GF07614468
Polycarbonate,
(PC), film, thickness 0.75 mm, size 100 × 100 mm, clear
Expand
View Pricing
GF02721710
Polycarbonate,
PC - 30% Carbon Fiber Reinforced, granule, 3 mm nominal granule size, weight 200 g, black
Expand
View Pricing
Page 1 of 6
Page 1 of 6