Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Platinum-rhodium alloy, Pt87Rh13
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 25
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF34358306
Platinum-rhodium alloy, Pt87Rh13,
2m wire, 0.125mm diameter, annealed
Expand
View Pricing
GF91057125
Platinum-rhodium alloy, Pt87Rh13,
1m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF67585225
Platinum-rhodium alloy, Pt87Rh13,
0.2m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF43007352
Platinum-rhodium alloy, Pt87Rh13,
20m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF82140353
Platinum-rhodium alloy, Pt87Rh13,
5m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF09528453
Platinum-rhodium alloy, Pt87Rh13,
0.5m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF84076229
Platinum-rhodium alloy, Pt87Rh13,
0.1m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF21838686
Platinum-rhodium alloy, Pt87Rh13,
10m wire, 0.025mm diameter, annealed
Expand
View Pricing
GF85774708
Platinum-rhodium alloy, Pt87Rh13,
5m wire, 0.025mm diameter, annealed
Expand
View Pricing
GF19090885
Platinum-rhodium alloy, Pt87Rh13,
1m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF48152729
Platinum-rhodium alloy, Pt87Rh13,
0.5m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF97250340
Platinum-rhodium alloy, Pt87Rh13,
0.1m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF67544051
Platinum-rhodium alloy, Pt87Rh13,
10m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF84686192
Platinum-rhodium alloy, Pt87Rh13,
1m wire, 0.025mm diameter, annealed
Expand
View Pricing
GF87877489
Platinum-rhodium alloy, Pt87Rh13,
0.5m wire, 0.025mm diameter, annealed
Expand
View Pricing
GF88261296
Platinum-rhodium alloy, Pt87Rh13,
0.2m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF91813642
Platinum-rhodium alloy, Pt87Rh13,
0.05m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF86361071
Platinum-rhodium alloy, Pt87Rh13,
2m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF29472600
Platinum-rhodium alloy, Pt87Rh13,
100m wire, 0.025mm diameter, annealed
Expand
View Pricing
GF29835782
Platinum-rhodium alloy, Pt87Rh13,
50m wire, 0.025mm diameter, annealed
Expand
View Pricing
Page 1 of 2
Page 1 of 2