Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Platinum-rhodium alloy, Pt70Rh30
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
GF38659359
Platinum-rhodium alloy, Pt70Rh30,
1m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF32419177
Platinum-rhodium alloy, Pt70Rh30,
2m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF73269928
Platinum-rhodium alloy, Pt70Rh30,
1m wire, 0.10mm diameter, annealed
Expand
View Pricing
GF88208286
Platinum-rhodium alloy, Pt70Rh30,
0.2m wire, 0.10mm diameter, annealed
Expand
View Pricing
GF24932325
Platinum-rhodium alloy, Pt70Rh30,
0.5m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF49072563
Platinum-rhodium alloy, Pt70Rh30,
0.5m wire, 0.10mm diameter, annealed
Expand
View Pricing
GF01106323
Platinum-rhodium alloy, Pt70Rh30,
0.05m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF72008153
Platinum-rhodium alloy, Pt70Rh30,
0.2m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF34071937
Platinum-rhodium alloy, Pt70Rh30,
0.1m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF43556834
Platinum-rhodium alloy, Pt70Rh30,
2m wire, 0.10mm diameter, annealed
Expand
View Pricing
GF61909274
Platinum-rhodium alloy, Pt70Rh30,
0.5m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF98095270
Platinum-rhodium alloy, Pt70Rh30,
0.2m wire, 0.25mm diameter, annealed
Expand
View Pricing