Skip to Content
Merck
Platinum octaethylporphyrin 95%

Platinum octaethylporphyrin

Platinum octaethylporphyrin 95%
Synonym(s):
Platinum(II) 2,3,7,8,12,13,17,18-octaethyl-21H,23H-porphyrin, PtOEP
Empirical Formula (Hill Notation):
C36H44N4Pt
CAS Number:
Molecular Weight:
727.84

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing