Skip to Content
Merck
Piracetam European Pharmacopoeia (EP) Reference Standard

Piracetam

Piracetam European Pharmacopoeia (EP) Reference Standard
Synonym(s):
2-(2-Oxopyrrolidino)acetamide, 2-Oxo-1-pyrrolidineacetamide
Empirical Formula (Hill Notation):
C6H10N2O2
CAS Number:
Molecular Weight:
142.16

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
API Family
Special Grade
Showing 1-3 of 3