Skip to Content
Merck
Piperacillin sodium salt analytical standard

Piperacillin sodium salt

Piperacillin sodium salt analytical standard
Empirical Formula (Hill Notation):
C23H26N5NaO7S
CAS Number:
Molecular Weight:
539.54
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing