Phosphomolybdic Acid Solution
Synonym(s):
Phosphomolybdic acid solution, Molybdophosphoric acid
Linear Formula:
H3[P(Mo3O10)4]
CAS Number:
Molecular Weight:
1825.25
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Reagent Type
Showing 1-4 of 4