![Phenylbutyrate Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/157/621/1f1010cf-9c46-492e-8ef2-f36c112a24bf/640/1f1010cf-9c46-492e-8ef2-f36c112a24bf.png)
Phenylbutyrate Related Compound A
![Phenylbutyrate Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/157/621/1f1010cf-9c46-492e-8ef2-f36c112a24bf/640/1f1010cf-9c46-492e-8ef2-f36c112a24bf.png)
Synonym(s):
3-Benzoylpropionic acid, 4-Oxo-4-phenylbutyric acid
Linear Formula:
C6H5COCH2CH2COOH
CAS Number:
Molecular Weight:
178.18
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2