
Pheniramine Impurity A

Synonym(s):
2-Benzylpyridine
Empirical Formula (Hill Notation):
C12H11N
CAS Number:
Molecular Weight:
169.22
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2