Pethoxamid
Synonym(s):
2-Chloro-N-(2-ethoxyethyl)-N-(2-methyl-1-phenyl-1-propenyl)acetamide
Empirical Formula (Hill Notation):
C16H22ClNO2
CAS Number:
Molecular Weight:
295.80
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing