Skip to Content
Merck
Pesticide Surrogate Spike Mix certified reference material, TraceCERT®, 200 μg/mL each component in acetone, ampule of 1 mL

Pesticide Surrogate Spike Mix

Pesticide Surrogate Spike Mix certified reference material, TraceCERT®, 200 μg/mL each component in acetone, ampule of 1 mL
Synonym(s):
Pesticide Surrogate Spike Mix

Products

Product Category
Brand
Product Line
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique