![Pepstatin A microbial, ≥90% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/253/546/41bdeacc-9913-4ac3-96f3-726c5c85710a/640/41bdeacc-9913-4ac3-96f3-726c5c85710a.png)
Pepstatin A
![Pepstatin A microbial, ≥90% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/253/546/41bdeacc-9913-4ac3-96f3-726c5c85710a/640/41bdeacc-9913-4ac3-96f3-726c5c85710a.png)
Synonym(s):
InSolution Pepstatin A, Synthetic
Empirical Formula (Hill Notation):
C34H63N5O9
CAS Number:
Molecular Weight:
685.89
Products
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-4 of 4
Compare
Product Number
Description
Pricing