Pentadecylmagnesium bromide solution
Linear Formula:
CH3(CH2)14MgBr
CAS Number:
Molecular Weight:
315.62
Products
Brand
Boiling Point (°C)
Formula Weight
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing