
Penflufen

Synonym(s):
2′-[(RS)-1,3-Dimethylbutyl]-5-fluoro-1,3-dimethylpyrazole-4-carboxanilide, N-[2-(1,3-Dimethylbutyl)phenyl]-5-fluoro-1,3-dimethyl-1H-pyrazole-4-carboxamide
Empirical Formula (Hill Notation):
C18H24FN3O
CAS Number:
Molecular Weight:
317.40
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1