
Palladium(II) trifluoroacetate

Synonym(s):
Pd(TFA)2, Trifluoroacetic acid palladium(II) salt
Linear Formula:
(CF3COO)2Pd
CAS Number:
Molecular Weight:
332.45
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reaction Type
Reagent Type
Core
Showing 1-1 of 1