Skip to Content
Merck
N-Succinyl-Ala-Ala-Ala-p-nitroanilide elastase substrate

N-Succinyl-Ala-Ala-Ala-p-nitroanilide

N-Succinyl-Ala-Ala-Ala-p-nitroanilide elastase substrate
Synonym(s):
N-Succinyl-L-alanyl-L-alanyl-L-alanine 4-nitroanilide, N-Succinyl-tri-L-alanine 4-nitroanilide
Empirical Formula (Hill Notation):
C19H25N5O8
CAS Number:
Molecular Weight:
451.43
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1