Skip to Content
Merck
N-[(S)-(+)-1-(Ethoxycarbonyl)-3-phenylpropyl]-L-alanine 98%

N-[(S)-(+)-1-(Ethoxycarbonyl)-3-phenylpropyl]-L-alanine

N-[(S)-(+)-1-(Ethoxycarbonyl)-3-phenylpropyl]-L-alanine 98%
Linear Formula:
C6H5CH2CH2CH(CO2C2H5)NHCH(CH3)CO2H
CAS Number:
Molecular Weight:
279.33

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing