
Norbuprenorphine solution

Synonym(s):
Norbuprenorphine solution
Empirical Formula (Hill Notation):
C25H35NO4
CAS Number:
Molecular Weight:
413.55
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique