![Nonylphenol solution 1000 μg/mL in acetone, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/145/553/9bb19723-1dd5-4cf0-8a2a-4b41060f43b3/640/9bb19723-1dd5-4cf0-8a2a-4b41060f43b3.png)
Nonylphenol solution
![Nonylphenol solution 1000 μg/mL in acetone, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/145/553/9bb19723-1dd5-4cf0-8a2a-4b41060f43b3/640/9bb19723-1dd5-4cf0-8a2a-4b41060f43b3.png)
Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35
Products
Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing