Skip to Content
Merck
N,O-Bis(trimethylsilyl)trifluoroacetamide for GC derivatization, LiChropur™, ≥99.0%

N,O-Bis(trimethylsilyl)trifluoroacetamide

N,O-Bis(trimethylsilyl)trifluoroacetamide for GC derivatization, LiChropur™, ≥99.0%
Synonym(s):
BSTFA
Linear Formula:
CF3C[=NSi(CH3)3]OSi(CH3)3
CAS Number:
Molecular Weight:
257.40
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Reaction Type
Reagent Type
Technique