Skip to Content
Merck
N,N,N′,N′-Tetramethyl-p-phenylenediamine 99%, powder

N,N,N′,N′-Tetramethyl-p-phenylenediamine

N,N,N′,N′-Tetramethyl-p-phenylenediamine 99%, powder
Synonym(s):
TMPD, TMPDA, TMPPD, Wurster’s reagent
Linear Formula:
C6H4[N(CH3)2]2
Molecular Weight:
164.25
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing