Skip to Content
Merck
N-Nitrosopiperidine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

N-Nitrosopiperidine

N-Nitrosopiperidine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
1-Nitrosopiperidine, NPIP
Empirical Formula (Hill Notation):
C5H10N2O
CAS Number:
Molecular Weight:
114.15

Products

Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Technique