N-Nitroso-N-methylbutylamine
Synonym(s):
Methylbutylnitrosamine
Empirical Formula (Hill Notation):
C5H12N2O
CAS Number:
Molecular Weight:
116.16
Products
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing