![N,N′-Disuccinimidyl carbonate purum, ≥95.0% (NMR)](/deepweb/assets/sigmaaldrich/product/structures/403/331/0ddcf4e2-ed57-4fb0-9c98-5267feb9a558/640/0ddcf4e2-ed57-4fb0-9c98-5267feb9a558.png)
N,N′-Disuccinimidyl carbonate
![N,N′-Disuccinimidyl carbonate purum, ≥95.0% (NMR)](/deepweb/assets/sigmaaldrich/product/structures/403/331/0ddcf4e2-ed57-4fb0-9c98-5267feb9a558/640/0ddcf4e2-ed57-4fb0-9c98-5267feb9a558.png)
Synonym(s):
N-Succinimidyl carbonate, DSC, Di(N-succinimidyl) carbonate
Empirical Formula (Hill Notation):
C9H8N2O7
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reaction Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing