Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N
,
N
-Dimethylglycine
Synonym(s):
(Dimethylamino)acetic acid
Linear Formula:
(CH
3
)
2
NCH
2
COOH
CAS Number:
1118-68-9
Molecular Weight:
103.12
EC Number:
214-267-8
Products
Product Category
amino acids (1)
Brand
Sigma-Aldrich (1)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Amino Acid
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
D1156
N
,
N
-Dimethylglycine,
≥99%, suitable for ligand binding assays
Expand
View Pricing