Skip to Content
Merck
N-Methylpropargylamine 95%

N-Methylpropargylamine

N-Methylpropargylamine 95%
Synonym(s):
3-Methylamino-1-propyne
Linear Formula:
HC≡CCH2NHCH3
CAS Number:
Molecular Weight:
69.11
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing