N-Methyl-N-nitroso-2-phenylethylamine
Synonym(s):
N-Methyl-N-nitrosobenzeneethanamine, N-Methyl-N-nitrosophenethylamine, N-Nitroso-N-methyl-2-phenylethylamine, Nitroso-N-methyl-N-(2-phenyl)ethylamine
Empirical Formula (Hill Notation):
C9H12N2O
Molecular Weight:
164.20
Products
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2