![Niclosamide (anhydrous) European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/236/679/5220770e-f80a-45ec-b904-e27a7be5ed99/640/5220770e-f80a-45ec-b904-e27a7be5ed99.png)
Niclosamide (anhydrous)
![Niclosamide (anhydrous) European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/236/679/5220770e-f80a-45ec-b904-e27a7be5ed99/640/5220770e-f80a-45ec-b904-e27a7be5ed99.png)
Synonym(s):
Niclosamide, 2′,5-Dichloro-4′-nitrosalicylanilide
Empirical Formula (Hill Notation):
C13H8Cl2N2O4
CAS Number:
Molecular Weight:
327.12
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing