Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 21
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF70059821
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 25 x 25mm, 0.01mm thickness, light tight (lt)
Expand
View Pricing
GF94765865
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 100 x 100mm, 0.2mm thickness
Expand
View Pricing
GF20721923
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 4mm disks, 0.01mm thickness
Expand
View Pricing
GF32743380
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 15mm disks, 0.2mm thickness
Expand
View Pricing
GF11315337
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 50 x 50mm, 0.2mm thickness
Expand
View Pricing
GF57478449
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 50 x 50mm, 0.01mm thickness, light tight (lt)
Expand
View Pricing
GF24359564
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 10mm disks, 0.2mm thickness
Expand
View Pricing
GF12516825
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 50mm disks, 0.2mm thickness
Expand
View Pricing
GF65298156
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 8mm disks, 0.2mm thickness
Expand
View Pricing
GF90240229
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 6mm disks, 0.01mm thickness
Expand
View Pricing
GF03583436
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 10mm disks, 0.01mm thickness
Expand
View Pricing
GF48146227
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 25mm disks, 0.2mm thickness
Expand
View Pricing
GF25782936
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 6mm disks, 0.2mm thickness
Expand
View Pricing
GF83516171
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 4mm disks, 0.2mm thickness
Expand
View Pricing
GF67156117
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 0.1m coil, 0.2mm thickness
Expand
View Pricing
GF47928853
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 15mm disks, 0.01mm thickness
Expand
View Pricing
GF11050962
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 0.2m coil, 0.2mm thickness
Expand
View Pricing
GF80028660
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 8mm disks, 0.01mm thickness
Expand
View Pricing
GF91106513
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 25mm disks, 0.01mm thickness
Expand
View Pricing
GF76845550
Nickel-beryllium-titanium alloy, Ni97.5Be1.95Ti0.5,
foil, 1m coil, 0.2mm thickness
Expand
View Pricing
Page 1 of 2
Page 1 of 2