Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel aluminide
Products
Manufacturer
Physical Form
Particle Size (µm)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
GF63391293
Nickel aluminide,
powder, 150 max. part. size (micron), weight 500 g, 45 μm mean particle size
Expand
View Pricing
GF71628521
Nickel aluminide,
powder, 150 max. part. size (micron), weight 100 g, purity 99%
Expand
View Pricing
GF95722735
Nickel aluminide,
powder, 150 max. part. size (micron), purity 99%, weight 500 g
Expand
View Pricing
GF53565789
Nickel aluminide,
powder, max. particle size 150 micron, weight 200 g, purity 99%
Expand
View Pricing
GF10948417
Nickel aluminide,
powder, max. particle size 150 micron, weight 50 g, 45 μm mean particle size
Expand
View Pricing
GF26173021
Nickel aluminide,
powder, max. particle size 150 micron, weight 50 g, purity 99%
Expand
View Pricing
GF96397217
Nickel aluminide,
powder, 150 max. part. size (micron), weight 100 g, 45 μm mean particle size
Expand
View Pricing
GF48517050
Nickel aluminide,
powder, max. particle size 150 micron, weight 200 g, 45 μm mean particle size
Expand
View Pricing