
Neosolaniol

Synonym(s):
Neosolaniol from Fusarium sp., 4β,15-Diacetoxy-3α,8α-dihydroxy-12,13-epoxytrichothec-9-ene, 8-Hydroxydiacetoxyscirpenol, NEO
Empirical Formula (Hill Notation):
C19H26O8
CAS Number:
Molecular Weight:
382.40
Products
Product Category
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing