Skip to Content
Merck
N-Boc-indole-2-boronic acid ≥95%

N-Boc-indole-2-boronic acid

N-Boc-indole-2-boronic acid ≥95%
Synonym(s):
1-(tert-butoxycarbonyl)indole-2-boronic acid, N-(tert-butoxycarbonyl)indole-2-boronic acid
Empirical Formula (Hill Notation):
C13H16BNO4
CAS Number:
Molecular Weight:
261.08

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing