Nα-Tosyl-L-lysine chloromethyl ketone hydrochloride
Synonym(s):
(3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride, (3S)-7-Amino-1-chloro-3-tosylamino-2-heptanone hydrochloride, TLCK, Tosyl-L-lysyl-chloromethane hydrochloride
Empirical Formula (Hill Notation):
C14H21ClN2O3S · HCl
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-2 of 2
Compare
Product Number
Description
Pricing