Skip to Content
Merck
Naphthol AS-BI phosphate disodium salt hydrate ≥96.0% (HPLC)

Naphthol AS-BI phosphate disodium salt hydrate

Naphthol AS-BI phosphate disodium salt hydrate ≥96.0% (HPLC)
Synonym(s):
2-Naphthalenecarboxamide, 7-bromo-N-(2-methoxyphenyl)-3-(phosphonooxy)-, disodium salt hydrate
Empirical Formula (Hill Notation):
C18H13BrNNa2O6P · xH2O
CAS Number:
Molecular Weight:
496.16 (anhydrous basis)

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1