Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
closure for dropper Bottle
Products
Manufacturer
Diameter (mm)
Color
Package Size
Special Grade
Natural (1)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
TMO312760-0010
Nalgene
®
closure for dropper Bottle,
white polypropylene, diam. 16 mm, thread 15-415, case of 2000 ea
Expand
View Pricing
TMO312760-0040
Nalgene
®
closure for dropper Bottle,
green polypropylene, diam. 16 mm, thread 15-415, case of 2000 ea
Expand
View Pricing
TMO312760-0060
Nalgene
®
closure for dropper Bottle,
blue polypropylene, diam. 16 mm, thread 15-415, case of 2000 ea
Expand
View Pricing
TMO312760-0050
Nalgene
®
closure for dropper Bottle,
red polypropylene, diam. 16 mm, thread 15-415, case of 2000 ea
Expand
View Pricing
TMO312760-0020
Nalgene
®
closure for dropper Bottle,
yellow polypropylene, diam. 16 mm, thread 15-415, case of 2000 ea
Expand
View Pricing
TMO312760-0000
Nalgene
®
closure for dropper Bottle,
natural polypropylene, diam. 16 mm, thread 15-415, case of 2000 ea
Expand
View Pricing