Skip to Content
Merck
N6-Methyladenine ≥95%

N6-Methyladenine

N6-Methyladenine ≥95%
Synonym(s):
6-(Methylamino)purine, N-Methyladenine
Empirical Formula (Hill Notation):
C6H7N5
CAS Number:
Molecular Weight:
149.15

Products

Biological Source
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Research Area
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing