Skip to Content
Merck
vibrant-m

Mycophenolate Mofetil Related Compound A

vibrant-m
Synonym(s):
2-Morpholinoethyl (E)-6-(1,3-dihydro-4,6-dihydroxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-4-hexenoate
Empirical Formula (Hill Notation):
C22H29NO7
Molecular Weight:
419.47

Products

Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade