Skip to Content
Merck
vibrant-m

Multicomponent Alcohol Mix-4000

vibrant-m
Synonym(s):
Multi-component Alcohol Mix

Products

Product Category
Brand
Manufacturer
Physical Form
Format
Chemical Composition
Application
Package Size
Reference Material Type
Special Grade
Technique