![MTBSTFA (with 1% t-BDMCS) ampule of 5 × 1 mL, (with 1% t-BDMCS), analytical standard, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/379/928/ba8e791f-fc62-4cd3-a556-ca2b8e87ad30/640/ba8e791f-fc62-4cd3-a556-ca2b8e87ad30.png)
MTBSTFA
![MTBSTFA (with 1% t-BDMCS) ampule of 5 × 1 mL, (with 1% t-BDMCS), analytical standard, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/379/928/ba8e791f-fc62-4cd3-a556-ca2b8e87ad30/640/ba8e791f-fc62-4cd3-a556-ca2b8e87ad30.png)
Synonym(s):
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane, N-Methyl-N-tert-butyldimethylsilyltrifluoroacetamide, MTBSTFA + 1% TBDMSCl, Silylating mixture VI
Empirical Formula (Hill Notation):
C9H18F3NOSi
CAS Number:
Molecular Weight:
241.33
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reaction Type
Reagent Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing